4-tert-Butylbenzoic anhydride 95%

Supplier: Thermo Scientific Chemicals
Warning

H32422-03 H32422-06
AAH32422-03EA 71.8 USD
AAH32422-03 AAH32422-06
4-tert-Butylbenzoic anhydride 95%
4-tert-Butylbenzoic anhydride

Purity: 95% CAS No.: 22201-45-2 Molecular Formula: C22H26O3 Formula Weight: 338.45 Melting Point: 75-81° MDL No.: MFCD09757530

Formula: C₂₂H₂₆O₃
MW: 338.45 g/mol
Melting Pt: 75…81 °C
Storage Temperature: Ambient
MDL Number: MFCD09757530
CAS Number: 22201-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR