4-Fluoro-3-nitrobenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

H32391-03 H32391-06
AAH32391-03EA 33.6 USD
AAH32391-03 AAH32391-06
4-Fluoro-3-nitrobenzonitrile 97%
4-Fluoro-3-nitrobenzonitrile

Purity: 97% CAS No.: 1009-35-4 Molecular Formula: C7H3FN2O2 Formula Weight: 166.11 Physical Form: Solid Appearance: Cream to orange Melting Point: 90-94° UN No.: 3439 MDL No.: MFCD01632197

Formula: C₇H₃FN₂O₂
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01632197
CAS Number: 1009-35-4
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR