2-(tert-Butoxycarbonylamino)-1-ethanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol

H32356-06 H32356-14
AAH32356-06EA 58.7 USD
AAH32356-06 AAH32356-14
2-(tert-Butoxycarbonylamino)-1-ethanol 95%
2-(tert-Butoxycarbonylamino)-1-ethanol

Purity: 95% CAS No.: 26690-80-2 Molecular Formula: C7H15NO3 Formula Weight: 161.20 Refractive Index: 1.4490 Boiling Point: 92°/0.22mm MDL No.: MFCD00056657

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Boiling Pt: 92 °C/0.22 mm
Density: 1.042
Storage Temperature: Ambient
MDL Number: MFCD00056657
CAS Number: 26690-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR