2-Ethyl-5,5-dimethyl-1,3-dioxane 99%

Supplier: Thermo Scientific Chemicals
Warning

H32351-06 H32351-14
AAH32351-06EA 87.9 USD
AAH32351-06 AAH32351-14
2-Ethyl-5,5-dimethyl-1,3-dioxane 99%
2-Ethyl-5,5-dimethyl-1,3-dioxane

Purity: 99% CAS No.: 768-58-1 Molecular Formula: C8H16O2 Formula Weight: 144.21 Flash Point: 50°(122°F) Odor: Characteristic Boiling Point: 154° UN No.: 1993 MDL No.: MFCD16622268

Formula: C₈H₁₆O₂
Boiling Pt: 154 °C
Density: 0.88
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD16622268
CAS Number: 768-58-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR