Boc-DL-Pro-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid, N-Boc-DL-proline
H32347-03
H32347-06
AAH32347-03EA
60.6
USD
AAH32347-03
AAH32347-06
AAH32347-MD
Boc-DL-Pro-OH 98%
Boc-DL-Pro-OH
Purity: 98%
CAS No.: 59433-50-0
Molecular Formula: C10H17NO4
Formula Weight: 215.25
Melting Point: 101-104°
MDL No.: MFCD00520915
Formula:
C₁₀H₁₇NO₄ MW: 215.25 g/mol Melting Pt: 101…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00520915 CAS Number: 59433-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...