4-(Trifluoromethyl)phenylhydrazinium chloride 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 4-(Trifluoromethyl)phenylhydrazine hydrochloride, 4-Hydrazinobenzotrifluoride hydrochloride, 1-(4-(Trifluoromethyl)phenyl)hydrazine hydrochloride

H32340-03 H32340-09
AAH32340-03EA 36.4 USD
AAH32340-03 AAH32340-09
4-(Trifluoromethyl)phenylhydrazinium chloride 96%
4-(Trifluoromethyl)phenylhydrazinium chloride

Purity: 96% CAS No.: 2923-56-0 Molecular Formula: C7H7F3N2.HCl Formula Weight: 212.60 Melting Point: ca 210° dec. MDL No.: MFCD00204233

Formula: C₇H₈ClF₃N₂
Melting Pt: ∼ 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00204233
CAS Number: 2923-56-0
EINECS: 642-499-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR