2-Biphenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-([1,1'-Biphenyl]-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H32320-03 H32320-06
AAH32320-14EA 835.21 USD
AAH32320-14 AAH32320-03 AAH32320-06
2-Biphenylboronic acid pinacol ester 97%
2-Biphenylboronic acid pinacol ester

Purity: 97% CAS No.: 914675-52-8 Molecular Formula: C18H21BO2 Formula Weight: 280.17 Melting Point: 76-79° MDL No.: MFCD14636459

Formula: C₁₈H₂₁BO₂
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD14636459
CAS Number: 914675-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR