4-Methoxyanthranilic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4-methoxybenzoic acid

H32319-03 H32319-06
AAH32319-03EA 61.9 USD
AAH32319-03 AAH32319-06
4-Methoxyanthranilic acid 96%
4-Methoxyanthranilic acid

Purity: 96% CAS No.: 4294-95-5 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: ca 171° dec. MDL No.: MFCD00667729

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: ∼ 171 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00667729
CAS Number: 4294-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR