4-Bromo-o-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-2-methylphenol, 1-Bromo-4-hydroxy-3-methylbenzene

H32307-06 H32307-14
AAH32307-03EA 32.95 USD
AAH32307-03 AAH32307-06 AAH32307-14
4-Bromo-o-cresol 97%
4-Bromo-o-cresol

Purity: 97% CAS No.: 2362-12-1 Molecular Formula: C7H7BrO Formula Weight: 187.04 Odor: Phenol-like Melting Point: 62-66° MDL No.: MFCD00055435

Formula: C₇H₇BrO
MW: 187.04 g/mol
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00055435
CAS Number: 2362-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR