4-Chloropicolinic acid 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloropyridine-2-carboxylic acid

H32301-09 H32301-03
AAH32301-09EA 352 USD
AAH32301-09 AAH32301-03
4-Chloropicolinic acid 94%
4-Chloropicolinic acid

Purity: 94% CAS No.: 5470-22-4 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Melting Point: ca 180° dec. MDL No.: MFCD00191400

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: ca 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00191400
CAS Number: 5470-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR