2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Bromoacetyl)pyridine hydrobromide

H32285-03 H32285-06 H32285-14
AAH32285-03EA 81.4 USD
AAH32285-03 AAH32285-06 AAH32285-14
2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide 98%
2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide

Purity: 98% CAS No.: 5349-17-7 Molecular Formula: C7H6BrNO.HBr Formula Weight: 280.95 Odor: Weak Melting Point: ca 200° dec. UN No.: 3261 MDL No.: MFCD02681893

Formula: C₇H₇Br₂NO
MW: 280.95
Melting Pt: ca 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02681893
CAS Number: 5349-17-7
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR