2-Bromo-6-chlorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H32269-03 H32269-06
AAH32269-03EA 33.6 USD
AAH32269-03 AAH32269-06
2-Bromo-6-chlorobenzoic acid 98%
2-Bromo-6-chlorobenzoic acid

Purity: 98% CAS No.: 93224-85-2 Molecular Formula: C7H4BrClO2 Formula Weight: 235.47 Melting Point: 148-150° MDL No.: MFCD00672929

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00672929
CAS Number: 93224-85-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR