Dimethyl-2,6-pyridinedicarboxylate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dimethyl pyridine-2,6-dicarboxylate

H32251-09 H32251-18
AAH32251-09EA 58.6 USD
AAH32251-09 AAH32251-18
Dimethyl-2,6-pyridinedicarboxylate 98%
Dimethyl 2,6-Pyridinedicarboxylate

Purity: 98% CAS No.: 5453-67-8 Molecular Formula: C9H9NO4 Formula Weight: 195.17 Melting Point: 119-122° MDL No.: MFCD00134493

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00134493
CAS Number: 5453-67-8
EINECS: 226-697-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR