2,6-Difluoro-3-hydroxybenzaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32250-03
H32250-MD
AAH32250-03EA
114
USD
AAH32250-03
AAH32250-MD
2,6-Difluoro-3-hydroxybenzaldehyde 95%
2,6-Difluoro-3-hydroxybenzaldehyde
Purity: 95%
CAS No.: 152434-88-3
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Melting Point: 110-114°
MDL No.: MFCD08275284
Formula:
C₇H₄F₂O₂ Melting Pt: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275284 CAS Number: 152434-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...