2-Methylbenzo[b]thiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32224-03
H32224-06
AAH32224-03EA
101
USD
AAH32224-03
AAH32224-06
2-Methylbenzo[b]thiophene 98%
2-Methylbenzo[b]thiophene
Purity: 98%
CAS No.: 1195-14-8
Molecular Formula: C9H8S
Formula Weight: 148.23
Melting Point: 51-52°
MDL No.: MFCD00216250
Formula:
C₉H₈S Melting Pt: 51…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216250 CAS Number: 1195-14-8 EINECS: 214-792-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...