4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32208-03
H32208-06
AAH32208-03EA
89.8
USD
AAH32208-03
AAH32208-06
4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine 96%
4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine
Purity: 96%
CAS No.: 300816-22-2
Molecular Formula: C9H7ClN2S
Formula Weight: 210.69
Melting Point: 104-105°
MDL No.: MFCD00572400
Formula:
C₉H₇ClN₂S MW: 210.69 g/mol Melting Pt: 104…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00572400 CAS Number: 300816-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...