4-tert-Butyl-o-phenylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(tert-Butyl)benzene-1,2-diamine, 4-tert-Butylbenzene-1,2-diamine, 4-tert-Butyl-1,2-phenylenediamine, 1,2-Diamino-4-(dimethylethyl)benzene, 1,2-Diamino-4-tert-butylbenzene
H32205-03
H32205-06
AAH32205-03EA
55.7
USD
AAH32205-03
AAH32205-06
4-tert-Butyl-o-phenylenediamine 97%
4-tert-Butyl-o-phenylenediamine
Purity: 97%
CAS No.: 68176-57-8
Molecular Formula: C10H16N2
Formula Weight: 164.25
Melting Point: 98-100°
UN No.: 2811
MDL No.: MFCD00052695
Formula:
C₁₀H₁₆N₂ MW: 164.25 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052695 CAS Number: 68176-57-8 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...