2,2-Difluoro-4-pentenoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Difluoropent-4-enoic acid
H32173-03
H32173-06
AAH32173-03EA
64.2
USD
AAH32173-03
AAH32173-06
AAH32173-MD
2,2-Difluoro-4-pentenoic acid 97%
2,2-Difluoro-4-pentenoic acid
Purity: 97%
CAS No.: 55039-89-9
Molecular Formula: C5H6F2O2
Formula Weight: 136.10
Boiling Point: 56-58°/3mm
UN No.: 3265
MDL No.: MFCD09800642
Formula:
C₅H₆F₂O₂ MW: 136.1 g/mol Boiling Pt: 56…58 °C/3 mm Storage Temperature: Ambient |
MDL Number:
MFCD09800642 CAS Number: 55039-89-9 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...