4-Hydroxy-2-(trifluoromethyl)benzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

H32158-03 H32158-MD
AAH32158-03EA 114 USD
AAH32158-03 AAH32158-MD
4-Hydroxy-2-(trifluoromethyl)benzonitrile 98%
4-Hydroxy-2-(trifluoromethyl)benzonitrile

Purity: 98% CAS No.: 320-42-3 Molecular Formula: C8H4F3NO Formula Weight: 187.12 Melting Point: 120-124° MDL No.: MFCD06797340

Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD06797340
CAS Number: 320-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR