5-Bromo-2,4-difluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2,4-difluoro-5-methylbenzene

H32155-06 H32155-14
AAH32155-06EA 74.9 USD
AAH32155-06 AAH32155-14
5-Bromo-2,4-difluorotoluene 98%
5-Bromo-2,4-difluorotoluene

Purity: 98% CAS No.: 159277-47-1 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 MDL No.: MFCD15144620

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144620
CAS Number: 159277-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR