11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32150-03
H32150-06
AAH32150-03EA
114.79
USD
AAH32150-03
AAH32150-06
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide 95%
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide
Purity: 95%
CAS No.: 207399-07-3
Molecular Formula: C36H44ClIN2
Formula Weight: 667.12
Melting Point: 232-234°
MDL No.: MFCD00191907
Formula:
C₃₆H₄₄ClIN₂ Melting Pt: 232…234 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191907 CAS Number: 207399-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...