1-(2,4-Dimethylphenyl)ethanol 95%

Supplier: Thermo Scientific Chemicals

H32141-03 H32141-09
AAH32141-03EA 200 USD
AAH32141-03 AAH32141-09
1-(2,4-Dimethylphenyl)ethanol 95%
1-(2,4-Dimethylphenyl)ethanol

Purity: 95% CAS No.: 5379-19-1 Molecular Formula: C10H14O Formula Weight: 150.22 MDL No.: MFCD00060890

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00060890
CAS Number: 5379-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR