1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3,4-Tetrahydro-1-naphthoic acid

H32138-03 H32138-MD
AAH32138-03EA 211 USD
AAH32138-03 AAH32138-MD
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid

Purity: 98% CAS No.: 1914-65-4 Molecular Formula: C11H12O2 Formula Weight: 176.22 Melting Point: 82-85° MDL No.: MFCD02179127

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD02179127
CAS Number: 1914-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR