3-Chloro-5-fluorophenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H32137-03 H32137-06
AAH32137-03EA 137 USD
AAH32137-03 AAH32137-06
3-Chloro-5-fluorophenylboronic acid 95%
3-Chloro-5-fluorophenylboronic acid

Purity: 95% CAS No.: 328956-61-2 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 MDL No.: MFCD06801741

Formula: C₆H₅BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06801741
CAS Number: 328956-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR