Stains-all 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide, 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine

H32127-03 H32127-06
AAH32127-03EA 92 USD
AAH32127-03 AAH32127-06
Stains-all 96%
Stains-all

General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)


Purity: 96% CAS No.: 7423-31-6 Molecular Formula: C30H27BrN2S2 Formula Weight: 559.58 Beilstein Reference: 3865213 MDL No.: MFCD00078788

Formula: C₃₀H₂₇BrN₂S₂
MW: 559.584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078788
CAS Number: 7423-31-6
EINECS: 231-047-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR