3-Chloro-5-fluorobenzyl alcohol 98+%

Supplier: Thermo Scientific Chemicals
Warning

H32122-03 H32122-06
AAH32122-03EA 27.3 USD
AAH32122-03 AAH32122-06
3-Chloro-5-fluorobenzyl alcohol 98+%
3-Chloro-5-fluorobenzyl alcohol

Purity: 98+% CAS No.: 79944-64-2 Molecular Formula: C7H6ClFO Formula Weight: 160.58 Refractive Index: 1.5310

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788554
CAS Number: 79944-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR