2-Fluoro-4-methoxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

H32118-03 H32118-09
AAH32118-03EA 72.7 USD
AAH32118-03 AAH32118-09
2-Fluoro-4-methoxybenzyl alcohol 97%
2-Fluoro-4-methoxybenzyl alcohol

Purity: 97% CAS No.: 405-09-4 Molecular Formula: C8H9FO2 Formula Weight: 156.15 MDL No.: MFCD04116336

Formula: C₈H₉FO₂
MW: 156.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116336
CAS Number: 405-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR