Methyl-4,5-dibromo-2-thiophenecarboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4,5-dibromothiophene-2-carboxylate

H32113-03 H32113-06
AAH32113-03EA 84.4 USD
AAH32113-03 AAH32113-06
Methyl-4,5-dibromo-2-thiophenecarboxylate 97%
Methyl-4,5-dibromo-2-thiophenecarboxylate

Purity: 97% CAS No.: 62224-24-2 Molecular Formula: C6H4Br2O2S Formula Weight: 299.97 Melting Point: 77-81° MDL No.: MFCD00831688

Formula: C₆H₄Br₂O₂S
MW: 299.98 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00831688
CAS Number: 62224-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR