3-Bromo-4-heptanone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H32094-03
H32094-09
AAH32094-03EA
66.1
USD
AAH32094-03
AAH32094-09
3-Bromo-4-heptanone 98%
3-Bromo-4-heptanone
Purity: 98%
CAS No.: 42330-10-9
Molecular Formula: C7H13BrO
Formula Weight: 193.08
Flash Point: 75°(167°F)
Refractive Index: 1.4580
Boiling Point: 39°/1mm
UN No.: 3265
MDL No.: MFCD09757526
Formula:
C₇H₁₃BrO MW: 193.08 g/mol Boiling Pt: 39 °C/1 mm Density: 1.248 Flash Pt: 75 °C(167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD09757526 CAS Number: 42330-10-9 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...