3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32090-03
H32090-06
AAH32090-03EA
61.3
USD
AAH32090-03
AAH32090-06
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid 97%
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 97%
CAS No.: 1736-21-6
Molecular Formula: C11H8FNO3
Formula Weight: 221.19
Storage Temperature: 2°C to 8°C
Melting Point: 199-201°
MDL No.: MFCD03407356
Formula:
C₁₁H₈FNO₃ Melting Pt: 199…201 °C Storage Temperature: Ambient |
MDL Number:
MFCD03407356 CAS Number: 1736-21-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...