2,2-Difluorocyclopropanemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

H32083-03 H32083-MD
AAH32083-03EA 158 USD
AAH32083-03 AAH32083-MD
2,2-Difluorocyclopropanemethanol 97%
2,2-Difluorocyclopropanemethanol

Purity: 97% CAS No.: 509072-57-5 Molecular Formula: C4H6F2O Formula Weight: 108.09 Boiling Point: 129° UN No.: 1987 MDL No.: MFCD06797748

Formula: C₄H₆F₂O
MW: 108.09 g/mol
Boiling Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD06797748
CAS Number: 509072-57-5
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR