2,6-Difluoro-4-iodophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

H32080-03 H32080-09
AAH32080-03EA 57.6 USD
AAH32080-03 AAH32080-09
2,6-Difluoro-4-iodophenol 99%
2,6-Difluoro-4-iodophenol

Purity: 99% CAS No.: 950858-06-7 Molecular Formula: C6H3F2IO Formula Weight: 255.99 Melting Point: 70-73° MDL No.: MFCD09907951

Formula: C₆H₃F₂IO
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD09907951
CAS Number: 950858-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR