4-Bromo-2-pyridone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine
H32067-03
H32067-06
AAH32067-03EA
79.3
USD
AAH32067-03
AAH32067-06
AAH32067-MD
4-Bromo-2-pyridone 97%
4-Bromo-2-pyridone
Purity: 97%
CAS No.: 36953-37-4
Molecular Formula: C5H4BrNO
Formula Weight: 174.00
Melting Point: 201-204°
MDL No.: MFCD00234041
Formula:
C₅H₄BrNO Melting Pt: 201…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234041 CAS Number: 36953-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...