2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32059-03
H32059-06
AAH32059-03EA
70.3
USD
AAH32059-03
AAH32059-06
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carbonitrile
CAS No.: 70291-62-2
Molecular Formula: C8H8N2S
Formula Weight: 164.23
Physical Form: Crystalline solid
Appearance: Pale brown
Melting Point: 153-154°
UN No.: 3439
MDL No.: MFCD00121495
Formula:
C₈H₈N₂S Melting Pt: 153…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00121495 CAS Number: 70291-62-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...