6-Bromo-3-formylchromone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Bromo-4-oxo-4H-chromene-3-carbaldehyde
H32053-03
H32053-06
AAH32053-03EA
18.9
USD
AAH32053-03
AAH32053-06
6-Bromo-3-formylchromone 97%
6-Bromo-3-formylchromone
Purity: 97%
CAS No.: 52817-12-6
Molecular Formula: C10H5BrO3
Formula Weight: 253.05
Melting Point: 190-193°
MDL No.: MFCD00191849
Formula:
C₁₀H₅BrO₃ MW: 253.05 g/mol Melting Pt: 190…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191849 CAS Number: 52817-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...