3-Chloro-4-(trifluoromethyl)benzoic acid 97%, JRD
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chloro-α,α,α-trifluoro-p-toluic acid
H32038-03
H32038-06
AAH32038-03EA
38
USD
AAH32038-03
AAH32038-06
3-Chloro-4-(trifluoromethyl)benzoic acid 97%, JRD
3-Chloro-4-(trifluoromethyl)benzoic acid
Purity: 97%
CAS No.: 115754-20-6
Molecular Formula: C8H4ClF3O2
Formula Weight: 224.57
Melting Point: 158-161°
MDL No.: MFCD04972755
Formula:
C₈H₄ClF₃O₂ MW: 224.57 g/mol Melting Pt: 158…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972755 CAS Number: 115754-20-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...