2-Bromo-2'-(trifluoromethyl)acetophenone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[2-(trifluoromethyl)phenyl]-1-ethanone

H32032-03 H32032-06
AAH32032-03EA 47.2 USD
AAH32032-03 AAH32032-06 AAH32032-MD
2-Bromo-2'-(trifluoromethyl)acetophenone 97%
2-Bromo-2'-(trifluoromethyl)acetophenone

Purity: 97% CAS No.: 54109-16-9 Molecular Formula: C9H6BrF3O Formula Weight: 267.04 Melting Point: 24-32° MDL No.: MFCD03094304

Formula: C₉H₆BrF₃O
MW: 267.04 g/mol
Melting Pt: 24…32 °C
Storage Temperature: Ambient
MDL Number: MFCD03094304
CAS Number: 54109-16-9
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR