6-Amino-1-tetralone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H32028-03
H32028-09
AAH32028-03EA
34.7
USD
AAH32028-03
AAH32028-09
6-Amino-1-tetralone 97%
6-Amino-1-tetralone
Purity: 97%
CAS No.: 3470-53-9
Molecular Formula: C10H11NO
Formula Weight: 161.21
Storage Temperature: 2°C to 8°C
Melting Point: 128-130°
UN No.: 2811
MDL No.: MFCD00099462
Formula:
C₁₀H₁₁NO Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00099462 CAS Number: 3470-53-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...