2-Chloro-4-methoxybenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-p-anisic acid
H32014-03
H32014-06
AAH32014-03EA
70.2
USD
AAH32014-03
AAH32014-06
2-Chloro-4-methoxybenzoic acid 98%
2-Chloro-4-methoxybenzoic acid
Purity: 98%
CAS No.: 21971-21-1
Molecular Formula: C8H7ClO3
Formula Weight: 186.59
MDL No.: MFCD00085943
Formula:
C₈H₇ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00085943 CAS Number: 21971-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...