Cyclohexylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclohexaneacetic acid

H31986-06 H31986-14
AAH31986-06EA 44.2 USD
AAH31986-06 AAH31986-14
Cyclohexylacetic acid 98%
Cyclohexylacetic acid

Purity: 98% CAS No.: 5292-21-7 Molecular Formula: C8H14O2 Formula Weight: 142.2 Melting Point: 28-32° Boiling Point: 242-244° MDL No.: MFCD00001518

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 242…244 °C
Melting Pt: 28…32 °C
Density: 0.9985 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001518
CAS Number: 5292-21-7
EINECS: 226-132-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR