4-n-Dodecylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals

H31983-03 H31983-06
AAH31983-03EA 59.9 USD
AAH31983-03 AAH31983-06
4-n-Dodecylbenzoic acid 98%
4-n-Dodecylbenzoic acid

Purity: 98% CAS No.: 21021-55-6 Molecular Formula: C19H30O2 Formula Weight: 290.45 MDL No.: MFCD04972235

Formula: C₁₉H₃₀O₂
Storage Temperature: Ambient
MDL Number: MFCD04972235
CAS Number: 21021-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR