2-Bromo-3-fluoropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H31981-03
H31981-06
AAH31981-03EA
43.1
USD
AAH31981-03
AAH31981-06
2-Bromo-3-fluoropyridine 97%
2-Bromo-3-fluoropyridine
Purity: 97%
CAS No.: 40273-45-8
Molecular Formula: C5H3BrFN
Formula Weight: 175.99
Refractive Index: 1.5450
Formula:
C₅H₃BrFN Storage Temperature: Ambient |
MDL Number:
MFCD04114128 CAS Number: 40273-45-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...