2,6-Difluoro-4-hydroxybenzaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31974-03
H31974-MD
AAH31974-03EA
87.9
USD
AAH31974-03
AAH31974-MD
2,6-Difluoro-4-hydroxybenzaldehyde 95%
2,6-Difluoro-4-hydroxybenzaldehyde
Purity: 95%
CAS No.: 532967-21-8
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Melting Point: 179-182°
MDL No.: MFCD07772042
Formula:
C₇H₄F₂O₂ Melting Pt: 179…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD07772042 CAS Number: 532967-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...