1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-Methyl-3,5-bis(trifluoromethyl)benzyl alcohol, 1-(3,5-Bis(trifluoromethyl)phenyl)ethanol

H31972-03 H31972-06 H31972-14
AAH31972-03EA 29.2 USD
AAH31972-03 AAH31972-06 AAH31972-14
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 98%
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol

Purity: 98% CAS No.: 368-63-8 Molecular Formula: C10H8F6O Formula Weight: 258.16 Melting Point: 70-74° MDL No.: MFCD01632521

Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD01632521
CAS Number: 368-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR