2-Amino-3-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

H31970-03 H31970-06
AAH31970-03EA 73.8 USD
AAH31970-03 AAH31970-06
2-Amino-3-fluoropyridine 97%
2-Amino-3-fluoropyridine

Purity: 97% CAS No.: 21717-95-3 Molecular Formula: C5H5FN2 Formula Weight: 112.11 Odor: Characteristic Melting Point: 39-43° UN No.: 2811 MDL No.: MFCD04114135

Formula: C₅H₅FN₂
MW: 112.11 g/mol
Melting Pt: 39…43 °C
Storage Temperature: Ambient
MDL Number: MFCD04114135
CAS Number: 21717-95-3
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR