6-Bromo-2-chlorobenzothiazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31962-03
H31962-06
AAH31962-03EA
38.2
USD
AAH31962-03
AAH31962-06
6-Bromo-2-chlorobenzothiazole 97%
6-Bromo-2-chlorobenzothiazole
Purity: 97%
CAS No.: 80945-86-4
Molecular Formula: C7H3BrClNS
Formula Weight: 248.53
Melting Point: 98-100°
MDL No.: MFCD04971822
Formula:
C₇H₃BrClNS MW: 248.53 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD04971822 CAS Number: 80945-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...