6-Bromochromone-3-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H31905-03 H31905-06
AAH31905-03EA 64.3 USD
AAH31905-03 AAH31905-06
6-Bromochromone-3-carboxylic acid 97%
6-Bromochromone-3-carboxylic acid

Purity: 97% CAS No.: 51085-91-7 Molecular Formula: C10H5BrO4 Formula Weight: 269.05 Melting Point: 191-193° MDL No.: MFCD01548933

Formula: C₁₀H₅BrO₄
MW: 269.05 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD01548933
CAS Number: 51085-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR