2-Chloro-4-methylpyrimidine 99%

Supplier: Thermo Scientific Chemicals
Warning

H31903-03 H31903-09
AAH31903-03EA 20.1 USD
AAH31903-03 AAH31903-09
2-Chloro-4-methylpyrimidine 99%
2-Chloro-4-methylpyrimidine

Purity: 99% CAS No.: 13036-57-2 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Melting Point: 45-48° MDL No.: MFCD00054434

Formula: C₅H₅ClN₂
Melting Pt: 45…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00054434
CAS Number: 13036-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR