Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-Amino-4-phenylthiophene-3-carboxylate
H31867-03
H31867-09
AAH31867-03EA
25.5
USD
AAH31867-03
AAH31867-09
Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%
Ethyl-2-amino-4-phenylthiophene-3-carboxylate
Purity: 96%
CAS No.: 4815-36-5
Molecular Formula: C13H13NO2S
Formula Weight: 247.32
Physical Form: Crystalline solid
Appearance: Yellow
Melting Point: 90-94°
MDL No.: MFCD00126391
Formula:
C₁₃H₁₃NO₂S Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126391 CAS Number: 4815-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...