6-Aminopicolinic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Aminopyridine-2-carboxylic acid
H31861-09
H31861-03
AAH31861-09EA
295
USD
AAH31861-09
AAH31861-03
6-Aminopicolinic acid 95%
6-Aminopicolinic acid
Purity: 95%
CAS No.: 23628-31-1
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Odor: Odorless
Melting Point: ca 295° dec.
MDL No.: MFCD00233711
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol Melting Pt: ∼ 295 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00233711 CAS Number: 23628-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...